Assay Details
| Gene Symbol: CA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014116666.2 | XP_013972141.1 | 1508 | 155 | ||
| XM_022423127.1 | XP_022278835.1 | 1511 | 155 | |||
| XM_022423128.1 | XP_022278836.1 | 1245 | 155 | |||
More Information
Set Membership: |
|

CA10
Genomic Map