Assay Details
| Gene Symbol: ACTN4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616611.3 | XP_005616668.1 | 15 - 16 | 1963 | 112 | |
| XM_541640.6 | XP_541640.2 | 15 - 16 | 1963 | 112 | ||
| XM_848317.5 | XP_853410.1 | 15 - 16 | 1962 | 112 | ||
| XM_862275.5 | XP_867368.1 | 15 - 16 | 1963 | 112 | ||
| XM_862326.5 | XP_867419.1 | 15 - 16 | 1963 | 112 | ||
More Information
Set Membership: |
|

ACTN4
Genomic Map