Assay Details
| Gene Symbol: GJB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022415777.1 | XP_022271485.1 | 2 | 893 | 131 | |
| XM_022415778.1 | XP_022271486.1 | 2 | 908 | 131 | ||
| XM_538073.6 | XP_538073.1 | 2 | 910 | 131 | ||
| XM_852480.5 | XP_857573.1 | 2 | 1356 | 131 | ||
More Information
Set Membership: |
|

GJB1
Genomic Map