Assay Details
| Gene Symbol: ICAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003291.1 | NP_001003291.1 | 5 | 1166 | 124 | |
| GenBank mRNA | L31625.1 | - | 5 | 1145 | 124 | |
| M68524.1 | - | 3 | 596 | 124 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ICAM1
Genomic Map