Assay Details
| Gene Symbol: KCNA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313824.1 | NP_001300753.1 | 1 | 1439 | 120 | |
| XM_005637263.3 | XP_005637320.1 | 2 | 2516 | 120 | ||
More Information
Set Membership: |
|

KCNA1
Genomic Map