Assay Details
| Gene Symbol: KCNJ1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005619563.3 | XP_005619620.1 | 1338 | 194 | ||
| XM_014113290.2 | XP_013968765.1 | 1103 | 194 | |||
More Information
Set Membership: |
|

KCNJ1
Genomic Map