Assay Details
| Gene Symbol: POU1F1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001006949.1 | NP_001006950.1 | 842 | 146 | ||
| XM_022413060.1 | XP_022268768.1 | 711 | 146 | |||
| GenBank mRNA | AF035585.1 | - | 842 | 146 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

POU1F1
Genomic Map