Assay Details
| Gene Symbol: EGR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618922.3 | XP_005618979.1 | 3 | 1357 | 109 | |
| XM_022417463.1 | XP_022273171.1 | 7 | 1222 | 109 | ||
| XM_022417464.1 | XP_022273172.1 | 6 | 1395 | 109 | ||
| XM_022417465.1 | XP_022273173.1 | 7 | 1349 | 109 | ||
| XM_022417466.1 | XP_022273174.1 | 5 | 1800 | 109 | ||
| XM_022417467.1 | XP_022273175.1 | 4 | 1082 | 109 | ||
| XM_536361.6 | XP_536361.3 | 3 | 1000 | 109 | ||
More Information
Set Membership: |
|

EGR2
Genomic Map