Assay Details
| Gene Symbol: G6PD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005642040.3 | XP_005642097.1 | 10 - 11 | 1597 | 64 | |
| XM_538209.6 | XP_538209.2 | 10 - 11 | 1440 | 64 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

G6PD
Genomic Map