Assay Details
| Gene Symbol: SERPINC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_537187.7 | XP_537187.4 | 3 - 4 | 803 | 171 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SERPINC1| Gene Symbol: SERPINC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_537187.7 | XP_537187.4 | 3 - 4 | 803 | 171 | |
Set Membership: |
|