Assay Details
| Gene Symbol: ACHE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005621023.3 | XP_005621080.1 | 2522 | 94 | ||
| XM_014114202.2 | XP_013969677.1 | 2823 | 94 | |||
| XM_546946.6 | XP_546946.2 | 2519 | 94 | |||
More Information
Set Membership: |
|

ACHE
Genomic Map