Assay Details
| Gene Symbol: ADORA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003279.1 | NP_001003279.1 | 932 | 130 | ||
| XM_005622154.3 | XP_005622211.1 | 1021 | 130 | |||
| XM_005622155.3 | XP_005622212.1 | 980 | 130 | |||
| XM_022420738.1 | XP_022276446.1 | 1127 | 130 | |||
| GenBank mRNA | X14051.1 | - | 932 | 130 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ADORA1
Genomic Map