Assay Details
| Gene Symbol: ANXA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001286970.1 | NP_001273899.1 | 4 - 5 | 418 | 161 | |
| GenBank mRNA | DN380016.1 | - | 4 - 5 | 422 | 161 | |
| DN870932.1 | - | 4 - 5 | 338 | 161 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ANXA1
Genomic Map