Assay Details
| Gene Symbol: OXTR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198659.1 | NP_001185588.1 | 680 | 78 | ||
| XM_022406234.1 | XP_022261942.1 | 883 | 78 | |||
| XM_022406235.1 | XP_022261943.1 | 892 | 78 | |||
| XM_022406236.1 | XP_022261944.1 | 892 | 78 | |||
| XM_022406237.1 | XP_022261945.1 | 892 | 78 | |||
| GenBank mRNA | HM856596.1 | - | 680 | 78 | ||
More Information
Set Membership: |
|

OXTR
Genomic Map