Assay Details
| Gene Symbol: PLA2G5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003433752.4 | XP_003433800.1 | 5 | 1032 | 131 | |
| XM_005617874.3 | XP_005617931.1 | 3 | 500 | 131 | ||
| XM_005617875.3 | XP_005617932.1 | 4 | 490 | 131 | ||
| XM_014110941.2 | XP_013966416.1 | 4 | 776 | 131 | ||
| XM_014110945.2 | XP_013966420.1 | 5 | 783 | 131 | ||
| XM_014110954.2 | XP_013966429.1 | 5 | 747 | 131 | ||
| XM_014110967.2 | XP_013966442.1 | 4 | 526 | 131 | ||
| XM_022413847.1 | XP_022269555.1 | 4 | 538 | 131 | ||
| XM_022413880.1 | XP_022269588.1 | 5 | 539 | 131 | ||
| XM_022413884.1 | XP_022269592.1 | 4 | 428 | 131 | ||
| XM_022413888.1 | XP_022269596.1 | 6 | 843 | 131 | ||
| XM_535382.6 | XP_535382.3 | 4 | 548 | 131 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PLA2G5
Genomic Map