Assay Details
| Gene Symbol: CYB5R2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022407794.1 | XP_022263502.1 | 6 - 7 | 549 | 161 | |
| XM_542485.5 | XP_542485.4 | 7 - 8 | 766 | 161 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CYB5R2
Genomic Map