Assay Details
| Gene Symbol: ALKBH2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005636332.3 | XP_005636389.1 | 3 | 793 | 186 | |
| XM_005636333.3 | XP_005636390.1 | 3 | 574 | 186 | ||
| XM_014107877.2 | XP_013963352.1 | 3 | 770 | 186 | ||
| XM_014107878.2 | XP_013963353.1 | 3 | 572 | 186 | ||
| XM_022410544.1 | XP_022266252.1 | 3 | 726 | 186 | ||
| XM_022410546.1 | XP_022266254.1 | 3 | 1356 | 186 | ||
| XM_534719.6 | XP_534719.2 | 3 | 714 | 186 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ALKBH2
Genomic Map