Assay Details
| Gene Symbol: PPP2R2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637895.3 | XP_005637952.1 | 7 | 888 | 219 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PPP2R2D| Gene Symbol: PPP2R2D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005637895.3 | XP_005637952.1 | 7 | 888 | 219 | |
Set Membership: |
|