Assay Details
| Gene Symbol: ACP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005616604.3 | XP_005616661.1 | 11 - 12 | 1686 | 178 | |
| XM_541628.5 | XP_541628.2 | 11 - 12 | 1507 | 178 | ||
More Information
Set Membership: |
|

ACP7
Genomic Map