Assay Details
| Gene Symbol: AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003053.1 | NP_001003053.1 | 2282 | 78 | ||
| GenBank mRNA | AF197950.1 | - | 2282 | 78 | ||
| AY271347.1 | - | 5 - 6 | 2285 | 78 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AR
Genomic Map