Assay Details
| Gene Symbol: MEN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081508.2 | NP_001074977.1 | 5 - 6 | 929 | 99 | |
| XM_005631057.3 | XP_005631114.1 | 6 - 7 | 1140 | 99 | ||
| XM_005631058.3 | XP_005631115.1 | 6 - 7 | 1152 | 99 | ||
| XM_005631059.3 | XP_005631116.1 | 6 - 7 | 1034 | 99 | ||
| XM_005631060.3 | XP_005631117.1 | 6 - 7 | 1137 | 99 | ||
| XM_022405078.1 | XP_022260786.1 | 6 - 7 | 955 | 99 | ||
| GenBank mRNA | DQ366289.1 | - | 5 - 6 | 929 | 99 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MEN1
Genomic Map