Assay Details
| Gene Symbol: POR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001177805.1 | NP_001171276.1 | 1239 | 87 | ||
| XM_005620899.3 | XP_005620956.1 | 1374 | 87 | |||
| XM_005620900.3 | XP_005620957.1 | 1326 | 87 | |||
| XM_005620901.3 | XP_005620958.1 | 1354 | 87 | |||
| XM_005620902.3 | XP_005620959.1 | 1444 | 87 | |||
| XM_005620904.3 | XP_005620961.1 | 1417 | 87 | |||
| XM_005620905.3 | XP_005620962.1 | 1435 | 87 | |||
| XM_005620906.3 | XP_005620963.1 | 1313 | 87 | |||
| XM_022419487.1 | XP_022275195.1 | 1340 | 87 | |||
| XM_022419488.1 | XP_022275196.1 | 1398 | 87 | |||
| GenBank mRNA | GU827421.1 | - | 1239 | 87 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

POR
Genomic Map