Assay Details
| Gene Symbol: RPL23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003100.2 | NP_001003100.2 | 1 | 215 | 185 | |
| GenBank mRNA | AJ388521.1 | - | 1 | 106 | 185 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RPL23
Genomic Map