Assay Details
| Gene Symbol: SMAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626569.2 | XP_005626626.1 | 2 - 3 | 775 | 95 | |
| XM_005626570.2 | XP_005626627.1 | 3 - 4 | 956 | 95 | ||
| XM_538641.6 | XP_538641.1 | 2 - 3 | 706 | 95 | ||
More Information
Set Membership: |
|

SMAD5
Genomic Map