Assay Details
| Gene Symbol: AP3S1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005626328.3 | XP_005626385.1 | 1 - 2 | 359 | 147 | |
| XM_022425131.1 | XP_022280839.1 | 359 | 147 | |||
| XM_853642.5 | XP_858735.2 | 1 - 2 | 359 | 147 | ||
More Information
Set Membership: |
|

AP3S1
Genomic Map