Assay Details
| Gene Symbol: MAGED1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641363.3 | XP_005641420.1 | 8 - 9 | 1933 | 97 | |
| XM_005641364.3 | XP_005641421.1 | 9 - 10 | 1904 | 97 | ||
| XM_005641365.3 | XP_005641422.1 | 8 - 9 | 1906 | 97 | ||
| XM_022416093.1 | XP_022271801.1 | 7 - 8 | 1857 | 97 | ||
| XM_022416094.1 | XP_022271802.1 | 7 - 8 | 1820 | 97 | ||
| XM_022416095.1 | XP_022271803.1 | 9 - 10 | 1877 | 97 | ||
| XM_022416096.1 | XP_022271804.1 | 8 - 9 | 1948 | 97 | ||
| XM_538044.7 | XP_538044.4 | 7 - 8 | 2000 | 97 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MAGED1
Genomic Map