Assay Details
| Gene Symbol: GALE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638903.4 | XP_003638951.1 | 10 - 11 | 1046 | 128 | |
| XM_022413386.1 | XP_022269094.1 | 9 - 10 | 989 | 128 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GALE
Genomic Map