Assay Details
| Gene Symbol: HMGB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_846436.5 | XP_851529.2 | 1 | 221 | 262 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_846436.5 transcripts.
This assay is in the 5'UTR region of the XM_846436.5 transcripts.
Set Membership: |
|

HMGB4
Genomic Map