Assay Details
| Gene Symbol: SLC45A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037947.1 | NP_001033036.1 | 2 - 3 | 642 | 84 | |
| XM_005618882.3 | XP_005618939.1 | 2 - 3 | 2018 | 84 | ||
| GenBank mRNA | DQ302162.1 | - | 2 - 3 | 642 | 84 | |
| KM222821.1 | - | 2 - 3 | 610 | 84 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

SLC45A2
Genomic Map