Assay Details
| Gene Symbol: LSP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005631336.2 | XP_005631393.2 | 6 - 7 | 750 | 153 | |
| XM_005631337.2 | XP_005631394.2 | 6 - 7 | 750 | 153 | ||
| XM_005631338.2 | XP_005631395.1 | 6 - 7 | 783 | 153 | ||
| XM_005631339.3 | XP_005631396.1 | 6 - 7 | 1525 | 153 | ||
| XM_022405521.1 | XP_022261229.1 | 6 - 7 | 699 | 153 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

LSP1
Genomic Map