Assay Details
| Gene Symbol: ZBTB16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014113409.2 | XP_013968884.1 | 1573 | 117 | ||
| XM_014113410.2 | XP_013968885.1 | 1596 | 117 | |||
| XM_022418298.1 | XP_022274006.1 | 1700 | 117 | |||
More Information
Set Membership: |
|

ZBTB16
Genomic Map