Assay Details
| Gene Symbol: ADSS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022421151.1 | XP_022276859.1 | 8 - 9 | 721 | 151 | |
| XM_022421152.1 | XP_022276860.1 | 7 - 8 | 670 | 151 | ||
| XM_022421153.1 | XP_022276861.1 | 9 - 10 | 982 | 151 | ||
| XM_022421154.1 | XP_022276862.1 | 8 - 9 | 778 | 151 | ||
| XM_537220.6 | XP_537220.2 | 7 - 8 | 763 | 151 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ADSS2
Genomic Map