Assay Details
| Gene Symbol: BCL2L1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003072.1 | NP_001003072.1 | 1 | 449 | 62 | |
| XM_005634674.3 | XP_005634731.1 | 2 | 974 | 62 | ||
| XM_005634675.3 | XP_005634732.1 | 3 | 491 | 62 | ||
| XM_005634676.3 | XP_005634733.1 | 2 | 992 | 62 | ||
| XM_005634677.3 | XP_005634734.1 | 3 | 605 | 62 | ||
| XM_014106833.2 | XP_013962308.1 | 3 | 623 | 62 | ||
| XM_014106834.2 | XP_013962309.1 | 3 | 509 | 62 | ||
| XM_022408808.1 | XP_022264516.1 | 2 | 998 | 62 | ||
| GenBank mRNA | AB073983.1 | - | 1 | 449 | 62 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

BCL2L1
Genomic Map