Assay Details
| Gene Symbol: CX3CR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001284491.1 | NP_001271420.1 | 2 | 659 | 169 | |
| XM_005634265.3 | XP_005634322.1 | 2 | 735 | 169 | ||
| XM_014106481.2 | XP_013961956.1 | 2 | 806 | 169 | ||
| XM_022408386.1 | XP_022264094.1 | 2 | 697 | 169 | ||
| XM_022408387.1 | XP_022264095.1 | 2 | 1680 | 169 | ||
| GenBank mRNA | AB648940.1 | - | 2 | 661 | 169 | |
More Information
Set Membership: |
|

CX3CR1
Genomic Map