Assay Details
| Gene Symbol: DUSP9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641992.3 | XP_005642049.1 | 3 | 754 | 130 | |
| XM_014111969.2 | XP_013967444.1 | 3 | 909 | 130 | ||
| XM_022416045.1 | XP_022271753.1 | 3 | 1376 | 130 | ||
More Information
Set Membership: |
|

DUSP9
Genomic Map