Assay Details
| Gene Symbol: GPC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014111827.2 | XP_013967302.1 | 5 - 6 | 937 | 165 | |
| XM_549265.6 | XP_549265.2 | 5 - 6 | 1287 | 165 | ||
More Information
Set Membership: |
|

GPC4
Genomic Map