Assay Details
| Gene Symbol: HCRTR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003638937.4 | XP_003638985.1 | 7 | 1364 | 86 | |
| XM_022412329.1 | XP_022268037.1 | 6 | 1065 | 86 | ||
More Information
Set Membership: |
|

HCRTR1
Genomic Map