Assay Details
| Gene Symbol: HSBP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003639067.2 | XP_003639115.1 | 1 - 2 | 165 | 77 | |
| XM_005620651.1 | XP_005620708.1 | 1 - 2 | 165 | 77 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HSBP1
Genomic Map