Assay Details
| Gene Symbol: AKT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001012340.1 | NP_001012340.1 | 3 - 4 | 386 | 99 | |
| XM_014114070.2 | XP_013969545.1 | 6 - 7 | 826 | 99 | ||
| XM_014114072.2 | XP_013969547.1 | 6 - 7 | 766 | 99 | ||
| XM_014114076.2 | XP_013969551.1 | 6 - 7 | 804 | 99 | ||
| XM_014114081.2 | XP_013969556.1 | 6 - 7 | 790 | 99 | ||
| XM_014114083.2 | XP_013969558.1 | 5 - 6 | 697 | 99 | ||
| XM_014114088.2 | XP_013969563.1 | 6 - 7 | 825 | 99 | ||
| GenBank mRNA | AY833721.1 | - | 3 - 4 | 386 | 99 | |
| AY833722.1 | - | 3 - 4 | 386 | 99 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

AKT2
Genomic Map