Assay Details
| Gene Symbol: SHROOM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005641094.3 | XP_005641151.1 | 1 - 2 | 171 | 156 | |
| XM_005641095.3 | XP_005641152.1 | 1 - 2 | 171 | 156 | ||
More Information
Set Membership: |
|

SHROOM2
Genomic Map