Assay Details
| Gene Symbol: AQP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014115489.2 | XP_013970964.1 | 3 - 4 | 711 | 135 | |
| XM_850363.5 | XP_855456.2 | 3 - 4 | 730 | 135 | ||
More Information
Set Membership: |
|

AQP4
Genomic Map