Assay Details
| Gene Symbol: RHOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003273.3 | NP_001003273.2 | 4 | 411 | 89 | |
| XM_005632310.3 | XP_005632367.1 | 5 | 554 | 89 | ||
| GenBank mRNA | CN004448.1 | - | 4 | 374 | 89 | |
| DR102913.1 | - | 2 | 152 | 89 | ||
| X56391.1 | - | 164 | 89 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RHOA
Genomic Map