Assay Details
| Gene Symbol: HAPLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014112236.2 | XP_013967711.1 | 914 | 120 | ||
| XM_844391.5 | XP_849484.1 | 886 | 120 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: HAPLN1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_014112236.2 | XP_013967711.1 | 914 | 120 | ||
| XM_844391.5 | XP_849484.1 | 886 | 120 | |||
Set Membership: |
|