Assay Details
| Gene Symbol: F2R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005618186.3 | XP_005618243.1 | 1121 | 120 | ||
| XM_014112263.2 | XP_013967738.1 | 1076 | 120 | |||
| XM_014112264.2 | XP_013967739.1 | 1097 | 120 | |||
| XM_014112265.2 | XP_013967740.1 | 1666 | 120 | |||
More Information
Set Membership: |
|

F2R
Genomic Map