Assay Details
| Gene Symbol: FKBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022411532.1 | XP_022267240.1 | 8 - 9 | 1044 | 93 | |
| XM_534923.6 | XP_534923.1 | 8 - 9 | 1225 | 93 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

FKBP4
Genomic Map