Assay Details
| Gene Symbol: GCG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003044.1 | NP_001003044.1 | 3 | 306 | 131 | |
| XM_022414644.1 | XP_022270352.1 | 3 | 455 | 131 | ||
| GenBank mRNA | AF308439.1 | - | 3 | 306 | 131 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GCG
Genomic Map