Assay Details
| Gene Symbol: DLA-DOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313787.1 | NP_001300716.1 | 3 | 477 | 133 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DLA-DOA| Gene Symbol: DLA-DOA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313787.1 | NP_001300716.1 | 3 | 477 | 133 | |
Set Membership: |
|