Assay Details
| Gene Symbol: HSPA4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048016.1 | NP_001041481.1 | 2 - 3 | 389 | 84 | |
| XM_005626399.2 | XP_005626456.1 | 2 - 3 | 224 | 84 | ||
| XM_022424911.1 | XP_022280619.1 | 3 - 4 | 302 | 84 | ||
| GenBank mRNA | AY911512.1 | - | 2 - 3 | 389 | 84 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

HSPA4
Genomic Map