Assay Details
| Gene Symbol: ID3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003025.2 | NP_001003025.1 | 1 | 281 | 126 | |
| GenBank mRNA | AJ271644.1 | - | 1 | 293 | 126 | |
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ID3
Genomic Map