Assay Details
| Gene Symbol: KCNJ10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005640901.3 | XP_005640958.1 | 3 | 642 | 113 | |
| XM_014111261.2 | XP_013966736.1 | 3 | 568 | 113 | ||
| XM_545752.6 | XP_545752.3 | 2 | 466 | 113 | ||
| GenBank mRNA | KU642467.1 | - | 1 | 253 | 113 | |
More Information
Set Membership: |
|

KCNJ10
Genomic Map